Phthalonitril 95%

Synonyme: Benzol-1,2-dicarbonitril,1,2-Benzoldicarbonitril,1,2-Dicyanobenzol,1,2-Dicyanbenzol,Phthalsäuredinitril

BD147936-500G BD147936-100G BD147936-25G
BLDPBD147936-500GEA 79 EUR
BLDPBD147936-500G BLDPBD147936-100G BLDPBD147936-25G
Phthalonitril 95%
Phthalonitril

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₄N₂
Molecular Weight: 128,13 g/mol
Lagertemperatur: Ambient
MDL: MFCD00001771
CAS-Nummer: 91-15-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR