Phthalimidoacetaldehyddiethylacetal 98%

BLDPBD147932-500GEA 699 EUR
BLDPBD147932-500G BLDPBD147932-100G BLDPBD147932-5G BLDPBD147932-10G BLDPBD147932-25G
Phthalimidoacetaldehyddiethylacetal 98%
Phthalimidoacetaldehyddiethylacetal

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₄H₁₇NO₄
Molecular Weight: 263,30 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005901
CAS-Nummer: 78902-09-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR