(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)acetaldehyd 98%

Synonyme: N-(Formylmethyl)phthalimide,Phthalimidoacetaldehyde

BD147931-5G BD147931-10G BD147931-1G BD147931-25G BD147931-100G
BLDPBD147931-5GEA 18 EUR
BLDPBD147931-5G BLDPBD147931-10G BLDPBD147931-1G BLDPBD147931-25G BLDPBD147931-100G
(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)acetaldehyd 98%
N-(2-Oxoethyl)phthalimide

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₀H₇NO₃
Molecular Weight: 189,17 g/mol
Lagertemperatur: Freezer
MDL: MFCD00023080
CAS-Nummer: 2913-97-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR