Phthalsäure 98%

Synonyme: 1,2-Benzoldicarbonsäure

BD147920-100G BD147920-25G BD147920-500G BD147920-1000G
BLDPBD147920-100GEA 10 EUR
BLDPBD147920-100G BLDPBD147920-25G BLDPBD147920-500G BLDPBD147920-1000G
Phthalsäure 98%
Phthalsäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆O₄
Molecular Weight: 166,13 g/mol
Lagertemperatur: Ambient
MDL: MFCD00002467
CAS-Nummer: 88-99-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR