Phthalhydrazid 99+%

Synonyme: 2,3-Dihydrophthalazine-1,4-dione

BD147919-100G BD147919-25G BD147919-10G BD147919-500G BD147919-1000G
BLDPBD147919-100GEA 21 EUR
BLDPBD147919-100G BLDPBD147919-25G BLDPBD147919-10G BLDPBD147919-500G BLDPBD147919-1000G
Phthalhydrazid 99+%
Phthalhydrazid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆N₂O₂
Molecular Weight: 162,15 g/mol
Lagertemperatur: Ambient
MDL: MFCD00006888
CAS-Nummer: 1445-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR