Phthalamidsäure 98%

BLDPBD147910-10GEA 72 EUR
BLDPBD147910-10G BLDPBD147910-25G BLDPBD147910-1G BLDPBD147910-100G BLDPBD147910-5G
Phthalamidsäure 98%
Phthalamidsäure

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₇NO₃
Molecular Weight: 165,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00025476
CAS-Nummer: 88-97-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR