Ethyl N-[4-(trifluoromethyl)pyrimid-2-yl]piperidine-4-carboxylate 98%

Synonyme: Ethyl 1-(4-(trifluoromethyl)pyrimidin-2-yl)piperidine-4-carboxylate

BD14789-5G BD14789-250MG BD14789-100MG BD14789-1G
BLDPBD14789-5GEA 594 EUR
BLDPBD14789-5G BLDPBD14789-250MG BLDPBD14789-100MG BLDPBD14789-1G
Ethyl N-[4-(trifluoromethyl)pyrimid-2-yl]piperidine-4-carboxylate 98%
Ethyl N-[4-(trifluoromethyl)pyrimid-2-yl]piperidine-4-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₃H₁₆F₃N₃O₂
Molecular Weight: 303,28 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00661832
CAS-Nummer: 215654-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR