Sodium phosphonoformate hexahydrate 98%

BLDPBD147854-250MGEA 6 EUR
BLDPBD147854-250MG BLDPBD147854-1G
Sodium phosphonoformate hexahydrate 98%
Sodium phosphonoformate hexahydrate

Storage condition: Inert atmosphere, Room Temperature

Formel: Na₂O₃PCO₂Na·6H₂O
Molecular Weight: 300,08 g/mol (191,98 anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150176
CAS-Nummer: 34156-56-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR