Triallyl(phenyl)silane 95%

Synonyme: Phenyltriallylsilane

BD147754-250MG BD147754-5G BD147754-1G
BLDPBD147754-250MGEA 13 EUR
BLDPBD147754-250MG BLDPBD147754-5G BLDPBD147754-1G
Triallyl(phenyl)silane 95%
Triallyl(phenyl)silane

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₅H₂₀Si
Molecular Weight: 228,41 g/mol
Lagertemperatur: Ambient
MDL: MFCD00094138
CAS-Nummer: 2633-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR