4-Cyanphenylboronsäurepinakolester 97%

Synonyme: 4-Cyanobenzeneboronic acid pinacol ester,4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile,4-Cyanophenylboronic acid, pinacol ester

BD14775-10G BD14775-1G BD14775-250MG BD14775-100G BD14775-5G BD14775-25G
BLDPBD14775-10GEA 65 EUR
BLDPBD14775-10G BLDPBD14775-1G BLDPBD14775-250MG BLDPBD14775-100G BLDPBD14775-5G BLDPBD14775-25G
4-Cyanphenylboronsäurepinakolester 97%
4-Cyanphenylboronsäurepinakolester

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₃H₁₆BNO₂
Molecular Weight: 229,09 g/mol
Lagertemperatur: Ambient
MDL: MFCD03093897
CAS-Nummer: 171364-82-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR