1-Phenylisochinolin 98%

BD147678-250MG BD147678-25G BD147678-5G BD147678-1G
BLDPBD147678-250MGEA 17 EUR
BLDPBD147678-250MG BLDPBD147678-25G BLDPBD147678-5G BLDPBD147678-1G
1-Phenylisochinolin 98%
1-Phenylisoquinoline

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₅H₁₁N
Molecular Weight: 205,2545 g/mol
Lagertemperatur: Ambient
MDL: MFCD00093934
CAS-Nummer: 3297-72-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR