Phenyl-β-D-glucopyranosid 99%

Synonyme: Phenyl-β-D-glucosid

BD147640-1G BD147640-100G BD147640-25G BD147640-5G
BLDPBD147640-1GEA 13 EUR
BLDPBD147640-1G BLDPBD147640-100G BLDPBD147640-25G BLDPBD147640-5G
Phenyl-β-D-glucopyranosid 99%
Phenyl-β-D-glucopyranosid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₆O₆
Molecular Weight: 256,26 g/mol (anhy)
Lagertemperatur: Ambient
MDL: MFCD00064089
CAS-Nummer: 1464-44-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR