4-(Carboxymethyl)phenylboronsäurepinakolester 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD147618-1GEA 74 EUR
BLDPBD147618-1G BLDPBD147618-250MG BLDPBD147618-100MG BLDPBD147618-5G
4-(Carboxymethyl)phenylboronsäurepinakolester 97%
4-(Carboxymethyl)phenylboronsäurepinakolester

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₄H₁₉BO₄
Molecular Weight: 262,1093 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04038755
CAS-Nummer: 797755-07-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR