Diphenylether 99%

Synonyme: Phenoxybenzol,Diphenyloxid

BD147565-25G
BLDPBD147565-25GEA 6 EUR
BLDPBD147565-25G
Diphenylether 99%
Diphenylether

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₀O
Molecular Weight: 170,21 g/mol
Lagertemperatur: Ambient
MDL: MFCD00003034
CAS-Nummer: 101-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR