Phenylbenzoat 99%

Synonyme: Benzoesäurephenylester

BD147555-500G BD147555-5G BD147555-100G BD147555-25G
BLDPBD147555-500GEA 246 EUR
BLDPBD147555-500G BLDPBD147555-5G BLDPBD147555-100G BLDPBD147555-25G
Phenylbenzoat 99%
Phenylbenzoat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₁₀O₂
Molecular Weight: 198,22 g/mol
Lagertemperatur: Ambient
MDL: MFCD00003072
CAS-Nummer: 93-99-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR