(S)-3-Methyl-2-((phenoxycarbonyl)amino)butanoic acid 95%

BD147512-1G BD147512-250MG BD147512-5G
BLDPBD147512-1GEA 124 EUR
BLDPBD147512-1G BLDPBD147512-250MG BLDPBD147512-5G
(S)-3-Methyl-2-((phenoxycarbonyl)amino)butanoic acid 95%
(S)-3-Methyl-2-((phenoxycarbonyl)amino)butanoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₅NO₄
Molecular Weight: 237,25 g/mol
Lagertemperatur: Ambient
MDL: MFCD03840384
CAS-Nummer: 126147-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR