Pheniramine maleate 98%

Synonyme: N,N-Dimethyl-3-phenyl-3-(pyridin-2-yl)propan-1-amine maleate

BD147448-1G BD147448-5G BD147448-250MG BD147448-25G
BLDPBD147448-1GEA 18 EUR
BLDPBD147448-1G BLDPBD147448-5G BLDPBD147448-250MG BLDPBD147448-25G
Pheniramine maleate 98%
Pheniramine maleate
Formel: C₂₀H₂₄N₂O₄
Molecular Weight: 356,422 g/mol
MDL: MFCD00079250
CAS-Nummer: 132-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR