Phenaceturic acid 97%

Synonyme: 2-(2-Phenylacetamido)acetic acid,N-(Phenylacetyl)glycine

BD147381-1G BD147381-25G BD147381-10G BD147381-100G BD147381-250MG BD147381-5G
BLDPBD147381-1GEA 16 EUR
BLDPBD147381-1G BLDPBD147381-25G BLDPBD147381-10G BLDPBD147381-100G BLDPBD147381-250MG BLDPBD147381-5G
Phenaceturic acid 97%
Phenaceturic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₁NO₃
Molecular Weight: 193,20 g/mol
Lagertemperatur: Ambient
MDL: MFCD00021744
CAS-Nummer: 500-98-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR