Undecafluorohexanoic acid 98%

Synonyme: Perfluorohexanoic Acid,IPC-PFFA-6

BD147248-1G BD147248-25G BD147248-5G
BLDPBD147248-1GEA 14 EUR
BLDPBD147248-1G BLDPBD147248-25G BLDPBD147248-5G
Undecafluorohexanoic acid 98%
Undecafluorohexanoic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆HF₁₁O₂
Molecular Weight: 314,05 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00198040
CAS-Nummer: 307-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR