Perfluoro-3,6,9-trioxatridecanoic acid 97%

Synonyme: 2,2-Difluoro-2-(1,1,2,2-tetrafluoro-2-(1,1,2,2-tetrafluoro-2-(perfluorobutoxy)ethoxy)ethoxy)acetic acid

BD147203-25G BD147203-5G BD147203-1G
BLDPBD147203-25GEA 539 EUR
BLDPBD147203-25G BLDPBD147203-5G BLDPBD147203-1G
Perfluoro-3,6,9-trioxatridecanoic acid 97%
Perfluoro-3,6,9-trioxatridecanoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀HF₁₉O₅
Molecular Weight: 562,09 g/mol
Lagertemperatur: Ambient
MDL: MFCD03094135
CAS-Nummer: 330562-41-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR