Pentafluorophenyl Acetate 98%

Synonyme: Acetic acid Pentafluorophenyl esterPerfluorophenyl acetate

BLDPBD147034-1GEA 35 EUR
BLDPBD147034-1G BLDPBD147034-250MG BLDPBD147034-5G BLDPBD147034-100MG BLDPBD147034-25G
Pentafluorophenyl Acetate 98%
Pentafluorophenyl Acetate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₃F₅O₂
Molecular Weight: 226,10 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00015552
CAS-Nummer: 19220-93-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR