Pentaerythritol tetrabenzoate 97%

Synonyme: 2,2-Bis((benzoyloxy)methyl)propane-1,3-diyl dibenzoate

BD146986-5G BD146986-25G BD146986-100G BD146986-500G
BLDPBD146986-5GEA 10 EUR
BLDPBD146986-5G BLDPBD146986-25G BLDPBD146986-100G BLDPBD146986-500G
Pentaerythritol tetrabenzoate 97%
Pentaerythritol tetrabenzoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₃₃H₂₈O₈
Molecular Weight: 552,58 g/mol
Lagertemperatur: Ambient
MDL: MFCD00020676
CAS-Nummer: 4196-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR