trans-3-Penten-1-ol 98%

Synonyme: (E)-3-Penten-1-ol

BLDPBD146537-250MGEA 118 EUR
BLDPBD146537-250MG BLDPBD146537-100MG BLDPBD146537-1G
trans-3-Penten-1-ol 98%
trans-3-Penten-1-ol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₅H₁₀O
Molecular Weight: 86,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07636756
CAS-Nummer: 764-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR