2-Oxo-2-(3,4,5-trimethoxyphenyl)acetic acid 95%

BLDPBD146519-100MGEA 41 EUR
BLDPBD146519-100MG BLDPBD146519-1G BLDPBD146519-250MG
2-Oxo-2-(3,4,5-trimethoxyphenyl)acetic acid 95%
2-Oxo-2-(3,4,5-trimethoxyphenyl)acetic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₁H₁₂O₆
Molecular Weight: 240,209 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03844682
CAS-Nummer: 88755-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR