Oxamic hydrazide 98%

Synonyme: Semioxamazide,Amino-oxamide,2-Hydrazinyl-2-oxoacetamide

BD146425-5G BD146425-1G BD146425-25G
BLDPBD146425-5GEA 32 EUR
BLDPBD146425-5G BLDPBD146425-1G BLDPBD146425-25G
Oxamic hydrazide 98%
Oxamic hydrazide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₂H₅N₃O₂
Molecular Weight: 103,08 g/mol
Lagertemperatur: Ambient
MDL: MFCD00008004
CAS-Nummer: 515-96-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR