Oxamidsäure 95%

Synonyme: Oxalsäuremonoamid

BD146421-5G BD146421-25G BD146421-1G
BLDPBD146421-5GEA 26 EUR
BLDPBD146421-5G BLDPBD146421-25G BLDPBD146421-1G
Oxamidsäure 95%
Oxamidsäure

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₂H₃NO₃
Molecular Weight: 89,05 g/mol
Lagertemperatur: Ambient
MDL: MFCD00008006
CAS-Nummer: 471-47-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR