Olsalazine Sodium 97%

BLDPBD146199-25GEA 175 EUR
BLDPBD146199-25G BLDPBD146199-10G BLDPBD146199-1G BLDPBD146199-5G
Olsalazine Sodium 97%
Olsalazine Sodium

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₄H₈N₂Na₂O₆
Molecular Weight: 346,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013300
CAS-Nummer: 6054-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR