Oleylamin 70%

Synonyme: (Z)-1-Amino-9-octadecen,cis-1-Amino-9-octadecen

BD146179-25G
BLDPBD146179-25GEA 7 EUR
BLDPBD146179-25G
Oleylamin 70%
Oleylamin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₈H₃₇N
Molecular Weight: 267,5 g/mol
Lagertemperatur: Ambient
MDL: MFCD00066507
CAS-Nummer: 112-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR