n-Octylamine hydrochloride 98%

Synonyme: OACl,Octan-1-amine hydrochloride,n-Octylammonium chloride

BD146104-5G BD146104-1G BD146104-250MG
BLDPBD146104-5GEA 119 EUR
BLDPBD146104-5G BLDPBD146104-1G BLDPBD146104-250MG
n-Octylamine hydrochloride 98%
n-Octylamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₁₉N·HCl
Molecular Weight: 165,71 g/mol
Lagertemperatur: Ambient
MDL: MFCD00042019
CAS-Nummer: 142-95-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR