Octylbutyrat 98+%
|
|
BLDPBD146094-100GEA
35
EUR
BLDPBD146094-100G
BLDPBD146094-25G
BLDPBD146094-500G
BLDPBD146094-1000G
Octylbutyrat 98+%
Octylbutyrat
Storage condition: Sealed in dry, Room Temperature
Formel:
C₁₂H₂₄O₂ Molecular Weight: 200,32 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00048939 CAS-Nummer: 110-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...