4-Methylbiphenyl 98%

Synonyme: 4-Phenyltoluol

BD14607-5G BD14607-25G BD14607-10G BD14607-100G BD14607-500G
BLDPBD14607-5GEA 8 EUR
BLDPBD14607-5G BLDPBD14607-25G BLDPBD14607-10G BLDPBD14607-100G BLDPBD14607-500G
4-Methylbiphenyl 98%
4-Methylbiphenyl

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₁₂
Molecular Weight: 168,24 g/mol
Lagertemperatur: Ambient
MDL: MFCD00008544
CAS-Nummer: 644-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR