2-Methoxyphenothiazine 98%

BD14604-5G BD14604-25G
BLDPBD14604-5GEA 18 EUR
BLDPBD14604-5G BLDPBD14604-25G
2-Methoxyphenothiazine 98%
2-Methoxyphenothiazine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₃H₁₁NOS
Molecular Weight: 229,30 g/mol
Lagertemperatur: Ambient
MDL: MFCD00037838
CAS-Nummer: 1771-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR