Octafluoro-4,4'-biphenol 95%

Synonyme: Octafluorobiphenyl-4,4'-diol,2,2',3,3',5,5',6,6'-Octafluoro-[1,1'-biphenyl]-4,4'-diol,Octafluoro-4,4'-dihydroxybiphenyl

BD146025-1G BD146025-25G BD146025-250MG BD146025-100G BD146025-5G
BLDPBD146025-1GEA 63 EUR
BLDPBD146025-1G BLDPBD146025-25G BLDPBD146025-250MG BLDPBD146025-100G BLDPBD146025-5G
Octafluoro-4,4'-biphenol 95%
Octafluoro-4,4'-biphenol

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₂F₈O₂
Molecular Weight: 330,13 g/mol
Lagertemperatur: Ambient
MDL: MFCD00045743
CAS-Nummer: 2200-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR