Magnesiumstearat Mg (4-5%)

Synonyme: Stearinsäure Magnesiumsalz

BD145997-100G BD145997-25G BD145997-500G BD145997-1000G
BLDPBD145997-100GEA 7 EUR
BLDPBD145997-100G BLDPBD145997-25G BLDPBD145997-500G BLDPBD145997-1000G
Magnesiumstearat Mg (4-5%)
Magnesiumstearat

Storage condition: Inert atmosphere, Room Temperature

Formel: C₃₆H₇₀MgO₄
Molecular Weight: 591,25 g/mol
Lagertemperatur: Ambient
MDL: MFCD00036391
CAS-Nummer: 557-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR