O-Benzyl-L-serine methyl ester hydrochloride 97%

Synonyme: H-Ser(Bzl)-OMe.HCl,Methyl O-Benzyl-L-serinate Hydrochloride

BD145953-10G BD145953-1G BD145953-5G BD145953-100G BD145953-25G BD145953-250MG
BLDPBD145953-10GEA 114 EUR
BLDPBD145953-10G BLDPBD145953-1G BLDPBD145953-5G BLDPBD145953-100G BLDPBD145953-25G BLDPBD145953-250MG
O-Benzyl-L-serine methyl ester hydrochloride 97%
O-Benzyl-L-serine methyl ester hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₁H₁₅NO₃
Molecular Weight: 245,70 g/mol
Lagertemperatur: Ambient
MDL: MFCD00191312
CAS-Nummer: 19525-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR