2-(Chloromethyl)benzoyl chloride 95%

BD145879-250MG BD145879-100MG BD145879-1G
BLDPBD145879-250MGEA 9 EUR
BLDPBD145879-250MG BLDPBD145879-100MG BLDPBD145879-1G
2-(Chloromethyl)benzoyl chloride 95%
2-(Chloromethyl)benzoyl chloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₆Cl₂O
Molecular Weight: 189,04 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00152348
CAS-Nummer: 42908-86-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR