O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride 98%

Synonyme: O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride for GC derivatization,PFBHA.HCl

BD145864-1G BD145864-250MG BD145864-100MG BD145864-5G
BLDPBD145864-1GEA 82 EUR
BLDPBD145864-1G BLDPBD145864-250MG BLDPBD145864-100MG BLDPBD145864-5G
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride 98%
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₄F₅NO
Molecular Weight: 249,57 g/mol
Lagertemperatur: Ambient
MDL: MFCD00012953
CAS-Nummer: 57981-02-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR