Tetradecylphosphonsäure 98%

Synonyme: 1-Tetradecylphosphonic acid,n-Tetradecylphosphonic acid,TDPA,N-tetradecylphosphonicacid

BD145779-1000G BD145779-25G BD145779-500G BD145779-100G
BLDPBD145779-1000GEA 205 EUR
BLDPBD145779-1000G BLDPBD145779-25G BLDPBD145779-500G BLDPBD145779-100G
Tetradecylphosphonsäure 98%
Tetradecylphosphonic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₃₁O₃P
Molecular Weight: 278,37 g/mol
Lagertemperatur: Ambient
MDL: MFCD00015834
CAS-Nummer: 4671-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR