Propylamine hydrochloride 98%

Synonyme: Propylamine Hydrochloride,1-Aminopropane Hydrochloride

BD145632-100G BD145632-5G BD145632-10G BD145632-25G
BLDPBD145632-100GEA 237 EUR
BLDPBD145632-100G BLDPBD145632-5G BLDPBD145632-10G BLDPBD145632-25G
Propylamine hydrochloride 98%
Propylamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₃H₉N
Molecular Weight: 95,57 g/mol
Lagertemperatur: Ambient
MDL: MFCD00012911
CAS-Nummer: 556-53-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR