5-Nitro-2-(3-phenylpropylamino)benzoic acid 98%

Synonyme: 5-Nitro-2-((3-phenylpropyl)amino)benzoic acid,NPPB

BD145604-1G BD145604-250MG
BLDPBD145604-1GEA 420 EUR
BLDPBD145604-1G BLDPBD145604-250MG
5-Nitro-2-(3-phenylpropylamino)benzoic acid 98%
5-Nitro-2-(3-phenylpropylamino)benzoic acid

Storage condition: Keep in dark place, Sealed in dry, Store in freezer, under -20°C

Formel: C₁₆H₁₆N₂O₄
Molecular Weight: 300,314 g/mol
Lagertemperatur: Freezer
MDL: MFCD00153851
CAS-Nummer: 107254-86-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR