2-Cinnamoylthiophene 98%

Synonyme: Styryl 2-Thienyl Ketone,3-Phenyl-1-(thiophen-2-yl)prop-2-en-1-one,3-Phenyl-1-(2-thienyl)-2-propen-1-one

BD14552-250MG BD14552-1G BD14552-25G BD14552-5G
BLDPBD14552-250MGEA 11 EUR
BLDPBD14552-250MG BLDPBD14552-1G BLDPBD14552-25G BLDPBD14552-5G
2-Cinnamoylthiophene 98%
2-Cinnamoylthiophene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₃H₁₀OS
Molecular Weight: 214,28 g/mol
Lagertemperatur: Ambient
MDL: MFCD00059742
CAS-Nummer: 3988-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR