4-Phenylbenzoesäure 98%

Synonyme: 4-BiphenylcarbonsäureBiphenyl-4-carbonsäure

BLDPBD14539-500GEA 106 EUR
BLDPBD14539-500G BLDPBD14539-25G BLDPBD14539-10G BLDPBD14539-1000G BLDPBD14539-100G BLDPBD14539-5G
4-Phenylbenzoesäure 98%
4-Phenylbenzoesäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₁₀O₂
Molecular Weight: 198,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002553
CAS-Nummer: 92-92-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR