Octylphosphonsäure 98%

Synonyme: OPA,1-Octylphosphonic acid,n-Octylphosphonic acid

BD145353-5G BD145353-25G BD145353-1G
BLDPBD145353-5GEA 10 EUR
BLDPBD145353-5G BLDPBD145353-25G BLDPBD145353-1G
Octylphosphonsäure 98%
Octylphosphonic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₁₉O₃P
Molecular Weight: 194,21 g/mol
Lagertemperatur: Ambient
MDL: MFCD00015841
CAS-Nummer: 4724-48-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR