Methoxy(dimethyl)octadecylsilane 98%

Synonyme: Methoxydimethyl(octadecyl)silane

BD145305-1G BD145305-5G
BLDPBD145305-1GEA 22 EUR
BLDPBD145305-1G BLDPBD145305-5G
Methoxy(dimethyl)octadecylsilane 98%
Methoxy(dimethyl)octadecylsilane

Storage condition: Inert atmosphere, Room Temperature

Formel: C₂₁H₄₆OSi
Molecular Weight: 342,68 g/mol
Lagertemperatur: Ambient
MDL: MFCD00042920
CAS-Nummer: 71808-65-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR