4-Benzenesulphonylbenzoic acid 95%

BLDPBD14530-1GEA 137 EUR
BLDPBD14530-1G BLDPBD14530-5G BLDPBD14530-100MG BLDPBD14530-250MG
4-Benzenesulphonylbenzoic acid 95%
4-Benzenesulphonylbenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₁₀O₄S
Molecular Weight: 262,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00568775
CAS-Nummer: 5361-54-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR