3-(4-Morpholinylcarbonyl)phenylboronsäure 98%

Synonyme: (3-(morpholine-4-carbonyl)phenyl)boronic acid,3-(Morpholine-4-carbonyl)phenylboronic acid,3-(4-Morpholinylcarbonyl)benzeneboronic acid

BD145222-250MG BD145222-100MG BD145222-5G BD145222-1G
BLDPBD145222-250MGEA 37 EUR
BLDPBD145222-250MG BLDPBD145222-100MG BLDPBD145222-5G BLDPBD145222-1G
3-(4-Morpholinylcarbonyl)phenylboronsäure 98%
3-(4-Morpholinylcarbonyl)phenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₁H₁₄BNO₄
Molecular Weight: 235,05 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD04115687
CAS-Nummer: 723281-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR