3,4,5,6-Tetrafluoro-N-methylphthalimide 97%

Synonyme: 4,5,6,7-Tetrafluoro-2-methylisoindoline-1,3-dione

BD145207-100G BD145207-25G BD145207-1G BD145207-5G
BLDPBD145207-100GEA 1000 EUR
BLDPBD145207-100G BLDPBD145207-25G BLDPBD145207-1G BLDPBD145207-5G
3,4,5,6-Tetrafluoro-N-methylphthalimide 97%
3,4,5,6-Tetrafluoro-N-methylphthalimide

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₃F₄NO₂
Molecular Weight: 233,12 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD01631699
CAS-Nummer: 33795-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR