N-Methylsaccharin 95%

Synonyme: 2-Methylbenzo[d]isothiazol-3(2H)-one 1,1-dioxide

BD145199-1G BD145199-25G BD145199-10G BD145199-250MG BD145199-5G
BLDPBD145199-1GEA 29 EUR
BLDPBD145199-1G BLDPBD145199-25G BLDPBD145199-10G BLDPBD145199-250MG BLDPBD145199-5G
N-Methylsaccharin 95%
N-Methylsaccharin

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₇NO₃S
Molecular Weight: 197,21 g/mol
Lagertemperatur: Ambient
MDL: MFCD00059263
CAS-Nummer: 15448-99-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR