N-Methylnicotinamid 96%
|
|
BLDPBD145158-5GEA
42
EUR
BLDPBD145158-5G
BLDPBD145158-100G
BLDPBD145158-25G
BLDPBD145158-1G
N-Methylnicotinamid 96%
N-Methylnicotinamid
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₇H₈N₂O Molecular Weight: 136,1512 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006384 CAS-Nummer: 114-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...