N-Methylnaphthalen-2-amine 95%

BD145155-100MG BD145155-250MG
BLDPBD145155-100MGEA 53 EUR
BLDPBD145155-100MG BLDPBD145155-250MG
N-Methylnaphthalen-2-amine 95%
N-Methylnaphthalen-2-amine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₁H₁₁N
Molecular Weight: 157,21 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00962475
CAS-Nummer: 2216-67-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR